siRNA Details
Gene Symbol: LOC101929563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121602.1 | 26, 27 | 3964 | |||
GenBank mRNA | CR627240.1 | - | 26, 27 | 3952 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000640307.1 | - | 3952 | Chr. - - [-] on Build |
