siRNA Details
Gene Symbol: LINC00462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_051983.1 | 3 | 349 | |||
GenBank mRNA | BM472085.1 | - | 3 | 350 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000425350.1 | - | 2 | 288 | Chr. 13 - 48576974 - 48578088 [-] on Build GRCh38 |
