siRNA Details
Gene Symbol: LINC01451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135288.1 | 3 | 3389 | |||
GenBank mRNA | AY129027.1 | - | 3 | 3397 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000628780.1 | - | 3389 | Chr. - - [-] on Build |
