siRNA Details
Gene Symbol: LOC107546764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136406.1 | 1, 2 | 142 | |||
GenBank mRNA | BX097691.1 | - | 1, 2 | 40 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000565120.1 | - | 1, 2 | 40 | Chr. 17 - 43938363 - 43938959 [+] on Build GRCh38 |
