siRNA Details
Gene Symbol: LINC01600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131168.1 | 3 | 1579 | |||
GenBank mRNA | AK056496.1 | - | 3 | 1579 | ||
BC104007.1 | - | 1 | 1363 | 40024375 | ||
BC104008.1 | - | 1 | 1363 | 40024376 | ||
BC104009.1 | - | 1 | 1363 | 40024379 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000624527.2 | - | 1579 | Chr. - - [-] on Build |
