siRNA Details
Gene Symbol: LINC00550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038878.1 | 3 | 2697 | |||
GenBank mRNA | BC042673.1 | - | 3 | 2697 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000629293.1 | - | 2697 | Chr. - - [-] on Build |
