siRNA Details
Gene Symbol: MIR34AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132742.1 | 2 | 3652 | |||
GenBank mRNA | EF570048.1 | - | 2 | 3419 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000635687.1 | - | 3706 | Chr. - - [-] on Build |
