siRNA Details
Gene Symbol: LINC01242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046204.1 | 3 | 468 | |||
GenBank mRNA | BC022036.1 | - | 4 | 1184 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000629369.1 | - | 1179 | Chr. - - [-] on Build |
