siRNA Details
Gene Symbol: LINC01012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038292.1 | 2 | 189 | |||
NR_038293.1 | 2 | 513 | ||||
NR_038294.1 | 2 | 513 | ||||
GenBank mRNA | AL120625.1 | - | 2 | 514 | ||
BC035101.1 | - | 2 | 189 | |||
BI771077.1 | - | 2 | 500 |
