siRNA Details
Gene Symbol: LOC100652758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132345.1 | 2 | 506 | |||
NR_132346.1 | 2 | 227 | ||||
GenBank mRNA | AB593120.1 | - | 2 | 142 | ||
CN293193.1 | - | 2 | 227 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100652758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132345.1 | 2 | 506 | |||
NR_132346.1 | 2 | 227 | ||||
GenBank mRNA | AB593120.1 | - | 2 | 142 | ||
CN293193.1 | - | 2 | 227 |