siRNA Details
Gene Symbol: C18orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272093.2 | NP_001259022.2 | 2 | 1647 | ||
GenBank mRNA | AK126293.1 | - | 2 | 1647 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000622985.1 | - | 1647 | Chr. - - [-] on Build |
