siRNA Details
Gene Symbol: FAM3D-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134853.1 | 2 | 225 | |||
GenBank mRNA | BG215913.1 | - | 2 | 225 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB166559.1 | - | 2 | 341 | Chr. 3 - 58606964 - 58608247 [+] on Build GRCh38 |
