siRNA Details
Gene Symbol: LINC00629 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038998.1 | 3 | 502 | |||
GenBank mRNA | AK022994.1 | - | 2 | 204 | ||
KM886854.1 | - | 2 | 405 | |||
KM886855.1 | - | 2 | 405 | |||
KM886856.1 | - | 3 | 393 | |||
KM886857.1 | - | 3 | 393 |
