siRNA Details
Gene Symbol: LINC00520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026796.1 | 4 | 550 | |||
NR_026797.1 | 3 | 488 | ||||
GenBank mRNA | BU171486.1 | - | 4 | 550 | ||
BU178088.1 | - | 3 | 500 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026796.1 | 4 | 550 | |||
NR_026797.1 | 3 | 488 | ||||
GenBank mRNA | BU171486.1 | - | 4 | 550 | ||
BU178088.1 | - | 3 | 500 |