siRNA Details
Gene Symbol: HYMAI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002768.2 | 1 | 1980 | |||
GenBank mRNA | AF241534.1 | - | 1 | 1980 | ||
BC059359.1 | - | 1 | 1501 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HYMAI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002768.2 | 1 | 1980 | |||
GenBank mRNA | AF241534.1 | - | 1 | 1980 | ||
BC059359.1 | - | 1 | 1501 |