siRNA Details
Gene Symbol: LUADT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132442.1 | 1 | 38 | |||
GenBank mRNA | N35358.1 | - | 1 | 38 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LUADT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132442.1 | 1 | 38 | |||
GenBank mRNA | N35358.1 | - | 1 | 38 |