siRNA Details
Gene Symbol: SMIM2-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046843.1 | 3 | 437 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SMIM2-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046843.1 | 3 | 437 |