siRNA Details
Gene Symbol: LINC00977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033916.1 | 3 | 890 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000509893.2 | - | 3 | 900 | Chr. 8 - 129216467 - 129241250 [-] on Build GRCh38 |
