siRNA Details
Gene Symbol: SNHG18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045196.2 | 3 | 899 | |||
GenBank mRNA | BM796927.1 | - | 1 | 402 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000508179.1 | - | 3 | 899 | Chr. 5 - 9546200 - 9550609 [+] on Build GRCh38 |
