siRNA Details
Gene Symbol: C12orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242696.1 | NP_001229625.1 | 4 | 798 | ||
GenBank mRNA | BC038743.1 | - | 4 | 766 | ||
BC117348.1 | - | 4 | 648 | |||
BC126393.1 | - | 4 | 648 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000551894.5 | - | 4 | 766 | Chr. 12 - 52205584 - 52210823 [-] on Build GRCh38 |
