siRNA Details
Gene Symbol: LOC729083 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122070.1 | 2 | 886 | |||
GenBank mRNA | BC038366.1 | - | 2 | 886 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000432377.1 | - | 2 | 886 | Chr. 3 - 42770612 - 42773635 [+] on Build GRCh38 |
