siRNA Details
Gene Symbol: LOC101928021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136398.1 | 1 | 378 | |||
GenBank mRNA | BI462408.1 | - | 1 | 341 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000585193.1 | - | 1 | 317 | Chr. 17 - 67244837 - 67245806 [+] on Build GRCh38 |

More Information
Other Species Targeted

