siRNA Details
Gene Symbol: DLGAP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024101.1 | 3 | 212 | |||
GenBank mRNA | AK093095.1 | - | 3 | 212 | ||
BC137437.1 | - | 2 | 95 | |||
BX647607.1 | - | 1 | 2155 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DLGAP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024101.1 | 3 | 212 | |||
GenBank mRNA | AK093095.1 | - | 3 | 212 | ||
BC137437.1 | - | 2 | 95 | |||
BX647607.1 | - | 1 | 2155 |