siRNA Details
Gene Symbol: LINC00398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047012.1 | 1 | 353 | |||
GenBank mRNA | DB005052.1 | - | 1 | 317 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047012.1 | 1 | 353 | |||
GenBank mRNA | DB005052.1 | - | 1 | 317 |