siRNA Details
Gene Symbol: LINC01578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037601.1 | 2 | 439 | |||
GenBank mRNA | AA083786.1 | - | 2 | 119 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000553829.1 | - | 2 | 550 | Chr. 15 - 92882732 - 92883861 [+] on Build GRCh38 |
