siRNA Details
Gene Symbol: TBX5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038440.1 | 3 | 514 | |||
GenBank mRNA | AK091766.1 | - | 2 | 497 | ||
CB992854.1 | - | 3 | 541 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TBX5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038440.1 | 3 | 514 | |||
GenBank mRNA | AK091766.1 | - | 2 | 497 | ||
CB992854.1 | - | 3 | 541 |