siRNA Details
Gene Symbol: LOC646268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033946.1 | 1 | 75 | |||
GenBank mRNA | BC029599.1 | - | 1 | 74 | ||
DB032996.1 | - | 1 | 75 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000414848.1 | - | 1 | 74 | Chr. 1 - 158132044 - 158140639 [-] on Build GRCh38 |
