siRNA Details
Gene Symbol: FLJ33581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040102.1 | 4 | 1736 | |||
GenBank mRNA | AK090900.1 | - | 4 | 1736 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000457274.1 | - | 4 | 1736 | Chr. 20 - 24199767 - 24224588 [+] on Build GRCh38 |
