siRNA Details
Gene Symbol: LINC01527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC042056.1 | - | 2 | 1090 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000419578.1 | - | 2 | 1090 | Chr. 1 - 152930040 - 152949210 [-] on Build GRCh38 |
