siRNA Details
Gene Symbol: LINC00927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033833.1 | 1 | 109 | |||
GenBank mRNA | AK097782.1 | - | 80 | |||
BC040586.1 | - | 1 | 72 | |||
DB448280.1 | - | 1 | 109 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033833.1 | 1 | 109 | |||
GenBank mRNA | AK097782.1 | - | 80 | |||
BC040586.1 | - | 1 | 72 | |||
DB448280.1 | - | 1 | 109 |