siRNA Details
Gene Symbol: C22orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302819.1 | NP_001289748.1 | 2 | 554 | ||
NM_001302820.1 | NP_001289749.1 | 2 | 481 | |||
NM_015372.2 | NP_056187.1 | 3 | 345 | |||
NR_126507.1 | 3 | 272 | ||||
GenBank mRNA | AJ704765.1 | - | 3 | 296 | ||
AJ704766.1 | - | 3 | 223 | |||
AK294232.1 | - | 2 | 524 | |||
AK315264.1 | - | 3 | 343 | |||
AL050256.1 | - | 3 | 333 | |||
DA485290.1 | - | 2 | 482 | |||
DB102820.1 | - | 3 | 272 |
