siRNA Details
Gene Symbol: HOXB-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033202.2 | 3 | 335 | |||
NR_033203.1 | 3 | 238 | ||||
NR_033205.1 | 3 | 331 | ||||
NR_110329.1 | 2 | 135 | ||||
NR_110330.1 | 2 | 232 | ||||
NR_110331.1 | 4 | 571 | ||||
GenBank mRNA | BG196429.1 | - | 2 | 140 | ||
BG203124.1 | - | 3 | 366 | |||
BG203609.1 | - | 3 | 361 | |||
BG217847.1 | - | 2 | 229 | |||
CB046509.1 | - | 3 | 247 |
