siRNA Details
Gene Symbol: LINC01116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040001.2 | 3 | 437 | |||
GenBank mRNA | BC013438.1 | - | 3 | 794 | ||
BC013588.1 | - | 1 | 106 | |||
BE908176.1 | - | 3 | 411 | |||
BF029957.1 | - | 3 | 433 |
