siRNA Details
Gene Symbol: Vmn1r52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053222.1 | NP_444452.1 | 1 | 721 |
Gene Symbol: V1ra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053223.1 | NP_444453.1 | 1 | 721 | ||
GenBank mRNA | BC138392.1 | - | 1 | 745 | 8861412 | |
BC138394.1 | - | 1 | 745 | 8861414 |
