siRNA Details
Gene Symbol: Vmn1r44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053227.2 | NP_444457.2 | 1 | 94 | ||
GenBank mRNA | BC107184.1 | - | 1 | 94 | ||
BC107185.1 | - | 1 | 94 | |||
BC151088.1 | - | 1 | 191 | 9088001 |
