siRNA Details
Gene Symbol: Vmn1r11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053233.2 | NP_444463.2 | 1 | 192 | ||
GenBank mRNA | BC104398.1 | - | 1 | 192 | 40050459 | |
BC104399.1 | - | 1 | 192 | 40050460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
