siRNA Details
Gene Symbol: Vmn1r25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053238.2 | NP_444468.2 | 1 | 454 |
Gene Symbol: Vmn1r30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134177.1 | NP_598938.1 | 1 | 454 | ||
GenBank mRNA | BC138075.1 | - | 1 | 503 | 8861094 | |
BC138078.1 | - | 1 | 503 | 8861097 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
