siRNA Details
Gene Symbol: Kiss1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053244.5 | NP_444474.1 | 1 | 1883 | ||
GenBank mRNA | AF343726.1 | - | 1 | 180 | ||
AK039628.1 | - | 1 | 1884 | |||
AK139303.1 | - | 1 | 1884 | |||
AY044228.1 | - | 1 | 216 | |||
BC016531.1 | - | 1 | 326 | 3488082 | ||
EU532436.1 | - | 1 | 180 | |||
EU532437.1 | - | 1 | 180 | |||
EU879091.1 | - | 1 | 180 | |||
GU075727.1 | - | 1 | 180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
