siRNA Details
Gene Symbol: Slc5a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053248.2 | NP_444478.2 | 13 | 1640 | ||
GenBank mRNA | AF235001.1 | - | 13 | 1607 | ||
AF380353.1 | - | 13 | 1562 | |||
AK046718.1 | - | 13 | 1641 | |||
BC137650.1 | - | 13 | 1575 | 8860669 | ||
BC137651.1 | - | 13 | 1575 | 8860670 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
