siRNA Details
Gene Symbol: Ehbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114595.1 | NP_001108067.1 | 11 | 1867 | ||
NM_001114596.1 | NP_001108068.1 | 12 | 1903 | |||
NM_001114597.1 | NP_001108069.1 | 12 | 4735 | |||
NM_053252.3 | NP_444482.2 | 12 | 2212 | |||
GenBank mRNA | AK087817.1 | - | 8 | 1454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
