siRNA Details
Gene Symbol: Ehbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114595.1 | NP_001108067.1 | 11 | 1863 | ||
NM_001114596.1 | NP_001108068.1 | 12 | 1899 | |||
NM_001114597.1 | NP_001108069.1 | 12 | 4731 | |||
NM_053252.3 | NP_444482.2 | 12 | 2208 | |||
GenBank mRNA | AF305087.1 | - | 12 | 4678 | ||
AF305088.1 | - | 12 | 2152 | |||
AF305089.1 | - | 11 | 1807 | |||
AF305090.1 | - | 12 | 1843 | |||
AK087817.1 | - | 8 | 1450 | |||
AK089917.1 | - | 11 | 1815 | |||
AK169974.1 | - | 12 | 2403 | |||
BC004660.1 | - | 12 | 2148 | 3497913 |
