siRNA Details
Gene Symbol: Prss28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053259.2 | NP_444489.2 | 4 | 513 | ||
GenBank mRNA | AF184895.1 | - | 3 | 352 | ||
AK144498.1 | - | 4 | 514 | |||
BC119386.1 | - | 3 | 380 | 8734135 | ||
BC119388.1 | - | 3 | 380 | 8734137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
