siRNA Details
Gene Symbol: Spred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033524.3 | NP_277059.1 | 6 | 1307 | ||
GenBank mRNA | AB063495.1 | - | 6 | 653 | ||
AK155065.1 | - | 6 | 1270 | |||
BC041685.1 | - | 5 | 621 | |||
BC079606.1 | - | 6 | 928 | 30355455 |
