siRNA Details
Gene Symbol: Spred2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033523.4 | NP_277058.1 | 6 | 1262 | ||
GenBank mRNA | AB063496.1 | - | 6 | 773 | ||
AK036430.1 | - | 6 | 1263 | |||
AK157817.1 | - | 6 | 1265 | |||
BC051247.1 | - | 1 | 206 | |||
BC066013.1 | - | 6 | 1332 | 5706237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
