siRNA Details
Gene Symbol: Pawr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054056.2 | NP_473397.1 | 7 | 1131 | ||
GenBank mRNA | AF377871.1 | - | 6 | 534 | ||
AK136412.1 | - | 2 | 96 | |||
DQ363525.1 | - | 7 | 1131 | |||
DQ449073.1 | - | 5 | 810 | |||
DQ449074.1 | - | 6 | 942 | |||
DV057979.1 | - | 2 | 114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
