siRNA Details
Gene Symbol: Psg28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054063.4 | NP_473404.3 | 2 | 412 | ||
GenBank mRNA | AF113598.1 | - | 2 | 262 | ||
BC099402.1 | - | 2 | 398 | 30787049 | ||
BC099403.1 | - | 2 | 283 | 30787292 | ||
BC141038.1 | - | 2 | 365 | 9055650 | ||
CK020635.1 | - | 2 | 412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
