siRNA Details
Gene Symbol: Dscaml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081270.1 | NP_001074739.1 | 5, 6 | 1122 | ||
GenBank mRNA | AF487345.1 | - | 5, 6 | 1026 | ||
AF487347.1 | - | 5, 6 | 1014 | |||
AK147531.1 | - | 5, 6 | 1066 | |||
AK161365.1 | - | 1151 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
