siRNA Details
Gene Symbol: Cygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030206.4 | NP_084482.1 | 2 | 749 | ||
GenBank mRNA | AJ315163.1 | - | 2 | 355 | ||
AK019410.1 | - | 2 | 750 | |||
AK087408.1 | - | 2 | 735 | |||
AK163073.1 | - | 2 | 500 | |||
AK172475.1 | - | 2 | 496 | |||
BC055040.1 | - | 4 | 658 | 6491041 |

More Information
Other Species Targeted

Gene Ontology Categories:
