siRNA Details
Gene Symbol: Vsx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054068.2 | NP_473409.1 | 5 | 1090 | ||
GenBank mRNA | AF391758.1 | - | 5 | 935 | ||
AF395732.1 | - | 5 | 1042 | |||
AK044393.1 | - | 5 | 973 | |||
AK044687.1 | - | 5 | 974 | |||
AK157311.1 | - | 5 | 1313 | |||
AY056825.1 | - | 5 | 955 | |||
BC057647.1 | - | 5 | 1090 | 4511665 | ||
BC138456.1 | - | 5 | 1020 | 8861476 | ||
BC138460.1 | - | 5 | 1020 | 8861480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
