siRNA Details
Gene Symbol: Fgfrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164259.1 | NP_001157731.1 | 5 | 1191 | ||
NM_054071.2 | NP_473412.1 | 6 | 1464 | |||
GenBank mRNA | AF321300.1 | - | 6 | 1191 | ||
AF321301.1 | - | 5 | 1152 | |||
AJ293947.1 | - | 5 | 1097 | |||
BC058745.1 | - | 5 | 1066 | 6439421 |
